Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024547.1 | 2 | 561 | |||
GenBank mRNA | DC364753.1 | - | 2 | 127 |
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298600.1 | - | 127 |