Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026717.1 | 2 | 446 | |||
GenBank mRNA | AK302111.1 | - | 2 | 460 | ||
BC110347.1 | - | 2 | 446 | 5165123 | ||
BC121128.1 | - | 1 | 280 | 40120738 | ||
BC121129.1 | - | 1 | 279 | 40120740 | ||
BC143833.1 | - | 1 | 316 | 9052348 | ||
BC143835.1 | - | 1 | 316 |