Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIPARP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027954.1 | 1 | 2102 | |||
GenBank mRNA | BC033324.1 | - | 1 | 2068 | ||
BC038770.1 | - | 1 | 736 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI460811.1 | - | 1 | 747 | Chr. 3 - 156673577 - 156674347 [-] on Build GRCh38 |