Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024546.1 | 3 | 469 | |||
XM_011526264.2 | XP_011524566.1 | 3 | 208 | |||
XM_017026094.1 | XP_016881583.1 | 3 | 208 | |||
GenBank mRNA | AK094926.1 | - | 4 | 1078 | ||
DA248363.1 | - | 3 | 208 |