Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027291.1 | 1 | 1586 | |||
GenBank mRNA | AK090915.1 | - | 1 | 1589 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM715105.1 | - | 1 | 224 | Chr. X - 73126037 - 73126718 [-] on Build GRCh38 |