Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033258.1 | 3 | 1125 | |||
GenBank mRNA | AF161345.1 | - | 1 | 526 | ||
AK124806.1 | - | 1 | 1184 |