Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028043.2 | 2 | 644 | |||
NR_133657.1 | 2 | 533 | ||||
GenBank mRNA | AB030733.1 | - | 2 | 637 | ||
AL521915.3 | - | 2 | 637 | |||
AL530868.3 | - | 2 | 538 | |||
AL545164.3 | - | 2 | 498 | |||
AY149295.1 | - | 2 | 512 | |||
AY149296.1 | - | 2 | 512 | |||
BC136585.1 | - | 2 | 517 | |||
BC142634.1 | - | 2 | 608 | |||
BC142691.1 | - | 2 | 608 |