Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002912.1 | 1 | 104 | |||
GenBank mRNA | Y11161.1 | - | 1 | 104 |
Gene Symbol: EIF4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296664.1 | - | 5 | 923 | ||
AK309274.1 | - | 7 | 1132 |