Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP6K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190316.1 | NP_001177245.1 | 3 | 1052 | ||
NM_001190317.1 | NP_001177246.1 | 3 | 975 | |||
NR_027437.2 | 3 | 574 | ||||
NR_027438.2 | 3 | 571 | ||||
XM_011533817.1 | XP_011532119.1 | 3 | 511 | |||
XM_011533818.1 | XP_011532120.1 | 2 | 779 | |||
XM_017006586.1 | XP_016862075.1 | 3 | 966 | |||
XM_017006587.1 | XP_016862076.1 | 4 | 701 | |||
XM_017006588.1 | XP_016862077.1 | 3 | 992 | |||
GenBank mRNA | AK097016.1 | - | 8 | 1694 | ||
AK290526.1 | - | 3 | 990 | |||
AK291501.1 | - | 3 | 967 | |||
AK304714.1 | - | 3 | 913 | |||
BC001864.1 | - | 3 | 496 | |||
BX537544.1 | - | 3 | 991 | |||
CD248129.1 | - | 3 | 537 |