Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027128.1 | 1 | 184 | |||
NR_027129.1 | 1 | 184 | ||||
GenBank mRNA | AF380178.1 | - | 1 | 116 | ||
AK127059.1 | - | 1 | 184 | |||
AY040088.1 | - | 1 | 116 | |||
BC128244.1 | - | 1 | 42 | |||
BC128245.1 | - | 1 | 42 | |||
BI829356.1 | - | 1 | 148 | |||
BI907880.1 | - | 1 | 184 |
Gene Symbol: FAM207A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG744869.1 | - | 6 | 757 |