Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002188.3 | 4 | 356 | |||
GenBank mRNA | AK295259.1 | - | 3 | 266 | ||
AK297653.1 | - | 3 | 241 | |||
BC000349.2 | - | 3 | 268 | |||
BC030240.1 | - | 4 | 235 | |||
BC092482.1 | - | 3 | 266 | |||
KJ690774.1 | - | 3 | 184 | |||
KJ690775.1 | - | 3 | 184 | |||
KJ690776.1 | - | 3 | 184 |