Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002188.3 | 9, 10 | 1073 | |||
GenBank mRNA | BC000349.2 | - | 7, 8 | 910 | ||
BC030240.1 | - | 9, 10 | 952 | |||
BC092482.1 | - | 7, 8 | 908 | |||
KJ690775.1 | - | 7, 8 | 826 | |||
KJ690776.1 | - | 7, 8 | 826 |
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KJ690772.1 | - | 8, 9 | 1075 |