Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027374.1 | 4 | 1082 | |||
GenBank mRNA | BC050337.1 | - | 4 | 1082 |