Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100101478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130172.1 | 4 | 723 | |||
GenBank mRNA | AY730279.1 | - | 4 | 723 | ||
BC028039.1 | - | 4 | 1369 |
Gene Symbol: H2BFXP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003238.2 | 4 | 723 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ217928.1 | - | 4 | 742 | Chr. X - 104060627 - 104062938 [-] on Build GRCh38 |