Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024271.1 | 11 | 1392 | |||
GenBank mRNA | AL832059.1 | - | 11 | 1255 | ||
BC092503.1 | - | 11 | 1337 |