Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024247.1 | 4 | 368 | |||
XM_011528385.2 | XP_011526687.1 | 5 | 490 | |||
GenBank mRNA | AK309092.1 | - | 3 | 187 | ||
AK310721.1 | - | 5 | 402 | |||
BC130443.1 | - | 4 | 328 | 40146474 | ||
BC144147.1 | - | 4 | 328 | |||
U20896.1 | - | 2 | 1297 |