Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR22HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028502.1 | 2 | 1946 | |||
NR_028503.1 | 3 | 1139 | ||||
NR_028504.1 | 4 | 726 | ||||
NR_028505.1 | 3 | 643 | ||||
GenBank mRNA | AF070569.1 | - | 1 | 1515 | ||
BX640650.1 | - | 4 | 664 | |||
BX648321.1 | - | 2 | 1883 |