Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHAMMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003521.1 | 7 | 1453 | |||
GenBank mRNA | BC048987.1 | - | 7 | 1513 | ||
BC093703.1 | - | 4 | 510 | |||
BC093705.1 | - | 4 | 510 | |||
DQ309040.1 | - | 7 | 1453 |
Gene Symbol: WHAMMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC117556.1 | - | 1077 |