Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHAMMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003521.1 | 2, 3 | 752 | |||
GenBank mRNA | AL833132.1 | - | 2, 3 | 363 | ||
BC018097.1 | - | 2, 3 | 230 | |||
BC044610.1 | - | 2, 3 | 363 | |||
BC048987.1 | - | 2, 3 | 812 | |||
DQ309039.1 | - | 2, 3 | 220 | |||
DQ309040.1 | - | 2, 3 | 752 |
Gene Symbol: WHAMMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC035099.1 | - | 2, 3 | 326 | ||
BC066982.1 | - | 2, 3 | 500 | |||
BC150524.1 | - | 1, 2 | 165 |