Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFEM1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021485.2 | 15 | 2324 | |||
GenBank mRNA | BC011266.2 | - | 5 | 620 | ||
BC103698.1 | - | 3 | 441 | |||
BC104748.1 | - | 3 | 440 | |||
BC105690.1 | - | 3 | 440 |