Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002963.1 | 1 | 12 | |||
GenBank mRNA | AJ609460.1 | - | 1 | 12 | ||
AJ609478.1 | - | 1 | 12 |