Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243079.1 | NP_001230008.1 | 14 | 1595 | ||
GenBank mRNA | AK054808.1 | - | 14 | 1595 | ||
AK122648.1 | - | 13 | 1767 | |||
BC100791.2 | - | 11 | 1159 | |||
BC100792.1 | - | 11 | 1159 | |||
DQ424898.1 | - | 7 | 768 |