Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027021.1 | 5 | 699 | |||
GenBank mRNA | AY077697.1 | - | 3 | 423 | ||
BG187681.1 | - | 5 | 704 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000423221.5 | - | 2 | 234 | Chr. 21 - 29748947 - 29764002 [+] on Build GRCh38 |