Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004390.1 | 1 | 126 |
Gene Symbol: METTL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005274232.4 | XP_005274289.1 | 1 | 296 |