Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023918.2 | 2 | 791 | |||
NR_023919.2 | 2 | 749 | ||||
NR_109759.1 | 2 | 745 | ||||
NR_109760.1 | 2 | 746 | ||||
NR_109761.1 | 3 | 866 | ||||
NR_109762.1 | 3 | 870 | ||||
GenBank mRNA | AF161506.1 | - | 2 | 720 | ||
BC016053.1 | - | 2 | 704 | |||
BC056896.1 | - | 1 | 349 | |||
CA489192.1 | - | 2 | 768 |