Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033169.1 | 2 | 251 | |||
XM_006715862.3 | XP_006715925.1 | 2 | 222 | |||
GenBank mRNA | AF440400.1 | - | 2 | 186 | ||
AK093091.1 | - | 2 | 204 | |||
AK315667.1 | - | 2 | 210 | |||
DB022678.1 | - | 2 | 98 |