Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026647.1 | 1 | 4582 | |||
GenBank mRNA | BC032951.1 | - | 2 | 3708 | ||
BC070120.1 | - | 2 | 3703 |