Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMBP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024130.2 | 5 | 1084 | |||
GenBank mRNA | AK098111.1 | - | 5 | 1086 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000434409.1 | - | 5 | 1084 | Chr. 10 - 99927209 - 99958381 [+] on Build GRCh38 |