Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024096.1 | 1 | 214 | |||
GenBank mRNA | AK094097.1 | - | 1 | 232 | ||
BX161445.1 | - | 1 | 214 |