Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003039.3 | 1, 2 | 281 | |||
GenBank mRNA | AJ489589.1 | - | 1, 2 | 248 |