Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002576.1 | 1 | 73 | |||
GenBank mRNA | AJ609436.1 | - | 1 | 73 | ||
AK095288.1 | - | 1 | 820 |