Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCG18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024052.2 | 5 | 2465 | |||
NR_024053.2 | 4 | 2251 | ||||
NR_102326.1 | 3 | 4122 | ||||
GenBank mRNA | AK055657.1 | - | 5 | 2159 | ||
AK056160.1 | - | 4 | 1468 | |||
AK098416.1 | - | 2 | 1001 |