Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU6F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026893.1 | 2 | 94 | |||
XM_017019507.1 | XP_016874996.1 | 2 | 1186 | |||
XM_017019508.1 | XP_016874997.1 | 3 | 1097 | |||
XM_017019509.1 | XP_016874998.1 | 1 | 106 | |||
XM_017019510.1 | XP_016874999.1 | 2 | 535 | |||
XM_017019511.1 | XP_016875000.1 | 2 | 704 | |||
XM_017019512.1 | XP_016875001.1 | 2 | 60 | |||
XM_017019513.1 | XP_016875002.1 | 2 | 1186 | |||
XM_017019514.1 | XP_016875003.1 | 2 | 1186 | |||
XM_017019515.1 | XP_016875004.1 | 2 | 532 | |||
XM_017019516.1 | XP_016875005.1 | 2 | 1186 | |||
XM_017019517.1 | XP_016875006.1 | 2 | 1186 | |||
XM_017019518.1 | XP_016875007.1 | 2 | 1186 | |||
XM_017019519.1 | XP_016875008.1 | 2 | 1186 | |||
XM_017019524.1 | XP_016875013.1 | 1 | 106 | |||
XM_017019525.1 | XP_016875014.1 | 2 | 94 | |||
XM_017019528.1 | XP_016875017.1 | 2 | 532 | |||
XM_017019529.1 | XP_016875018.1 | 2 | 532 | |||
GenBank mRNA | AI417271.1 | - | 2 | 156 | ||
AK095191.1 | - | 2 | 94 |