Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029608.1 | 1 | 94 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN286812.1 | - | 1 | 258 | Chr. 17 - 48579630 - 48580111 [-] on Build GRCh38 |