Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM41B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015012.3 | NP_055827.1 | 7 | 3685 | ||
NR_028491.2 | 8 | 1356 | ||||
GenBank mRNA | BC035034.1 | - | 7 | 3563 | 5259179 | |
BC044597.1 | - | 7 | 3553 | 4811565 | ||
BC091524.1 | - | 8 | 1155 | 30512350 |