Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS31P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002816.2 | 8 | 1819 | |||
NR_051963.1 | 8 | 1104 | ||||
NR_051964.1 | 6 | 689 | ||||
GenBank mRNA | AK027536.1 | - | 8 | 1819 | ||
AK307345.1 | - | 6 | 642 | |||
AK309380.1 | - | 8 | 1077 | |||
BX648914.1 | - | 8 | 1094 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA274275.1 | - | 5 | 461 | Chr. 13 - 52169278 - 52184102 [-] on Build GRCh38 |