Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARNA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003009.1 | 1 | 78 | |||
GenBank mRNA | AY077742.1 | - | 1 | 78 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CV569582.1 | - | 1 | 274 | Chr. 9 - 19063400 - 19063982 [-] on Build GRCh38 |