Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002985.2 | 1 | 121 | |||
GenBank mRNA | AJ609450.1 | - | 1 | 121 | ||
DW436402.1 | - | 1 | 137 |