Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002987.1 | 1 | 44 | |||
GenBank mRNA | AJ609451.1 | - | 1 | 44 |
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092096.1 | - | 2 | 1448 | ||
BC045792.1 | - | 4 | 638 | |||
BC064589.1 | - | 3 | 1169 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DR002536.1 | - | 4 | 614 | Chr. 1 - 28579688 - 28581865 [-] on Build GRCh38 |