Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003244.1 | 1 | 241 | |||
GenBank mRNA | DQ860409.1 | - | LIT3337 HAR1F |
1 | 241 |