Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003244.1 | 2 | 2702 | |||
GenBank mRNA | BC035016.1 | - | 2 | 1801 | ||
BC047717.1 | - | 2 | 1782 | |||
DQ860409.1 | - | LIT3337 HAR1F |
2 | 2702 |