Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D3P1-DHX40P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002924.3 | 5, 6 | 490 | |||
GenBank mRNA | BX280208.1 | - | 5, 6 | 413 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI828567.1 | - | 5, 6 | 413 | Chr. 17 - 59995951 - 60018975 [-] on Build GRCh38 |