Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028343.1 | 5 | 520 | |||
XM_006723267.1 | XP_006723330.1 | 4 | 452 | |||
XM_006723277.2 | XP_006723340.1 | 4 | 450 | |||
XM_006723279.1 | XP_006723342.1 | 4 | 609 | |||
XM_011527100.1 | XP_011525402.1 | 4 | 599 | |||
XM_011527101.2 | XP_011525403.1 | 4 | 574 | |||
XM_011527104.1 | XP_011525406.1 | 4 | 688 | |||
XM_017026952.1 | XP_016882441.1 | 4 | 402 | |||
XM_017026953.1 | XP_016882442.1 | 4 | 599 | |||
XM_017026954.1 | XP_016882443.1 | 4 | 476 | |||
XM_017026955.1 | XP_016882444.1 | 4 | 452 | |||
XM_017026956.1 | XP_016882445.1 | 5 | 602 | |||
XM_017026957.1 | XP_016882446.1 | 4 | 340 | |||
XM_017026958.1 | XP_016882447.1 | 5 | 656 | |||
XM_017026959.1 | XP_016882448.1 | 4 | 527 | |||
XM_017026960.1 | XP_016882449.1 | 3 | 242 | |||
XM_017026961.1 | XP_016882450.1 | 4 | 688 | |||
XM_017026962.1 | XP_016882451.1 | 5 | 631 | |||
XM_017026963.1 | XP_016882452.1 | 4 | 527 | |||
XM_017026968.1 | XP_016882457.1 | 5 | 715 | |||
XM_017026971.1 | XP_016882460.1 | 4 | 688 | |||
XM_017026974.1 | XP_016882463.1 | 4 | 566 | |||
GenBank mRNA | AK095504.1 | - | 5 | 492 | ||
BC063880.1 | - | 4 | 440 | 30388597 | ||
BC069668.1 | - | 4 | 439 | 7262632 | ||
DQ925698.1 | - | 4 | 540 |