Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR100HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024430.2 | 4 | 2660 | |||
NR_137175.1 | 2 | 2931 | ||||
NR_137176.1 | 2 | 2642 | ||||
NR_137177.1 | 2 | 2621 | ||||
NR_137179.1 | 5 | 2614 | ||||
NR_137180.1 | 5 | 2672 | ||||
NR_137181.1 | 3 | 2557 | ||||
NR_137182.1 | 4 | 2639 | ||||
NR_137183.1 | 3 | 2548 | ||||
NR_137184.1 | 2 | 2466 | ||||
NR_137185.1 | 3 | 2527 | ||||
NR_137186.1 | 3 | 2436 | ||||
NR_137187.1 | 3 | 2415 | ||||
NR_137188.1 | 3 | 2505 | ||||
NR_137189.1 | 2 | 2952 | ||||
NR_137190.1 | 3 | 2985 | ||||
NR_137191.1 | 3 | 2964 | ||||
NR_137192.1 | 4 | 2824 | ||||
NR_137193.1 | 5 | 2651 | ||||
NR_137194.1 | 3 | 2526 | ||||
NR_137195.1 | 2 | 2694 | ||||
NR_137196.1 | 2 | 2715 | ||||
GenBank mRNA | AB075498.1 | - | 2 | 3183 | ||
AK021552.1 | - | 1 | 1828 | |||
AK123947.1 | - | 2 | 2867 | |||
AL832469.1 | - | 3 | 2219 | |||
BC040599.1 | - | 2 | 2245 | |||
BX538058.1 | - | 2 | 2222 | |||
BX647608.1 | - | 2 | 2223 |