Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291641.1 | NP_001278570.1 | 2, 3 | 277 | ||
XM_017024387.1 | XP_016879876.1 | 2, 3 | 231 | |||
XM_017024388.1 | XP_016879877.1 | 2, 3 | 243 | |||
XM_017024392.1 | XP_016879881.1 | 2, 3 | 233 | |||
GenBank mRNA | AL833603.1 | - | 2, 3 | 217 | ||
HY030361.1 | - | 2, 3 | 276 |