Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002770.1 | 1 | 3357 | |||
GenBank mRNA | AF469201.1 | - | 2 | 2376 | ||
AF469202.1 | - | 1 | 3357 | |||
AF469203.1 | - | 2 | 888 | |||
AF469206.1 | - | 3 | 855 | |||
AF469207.1 | - | 3 | 762 | |||
AF469208.1 | - | 2 | 587 |