Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028089.1 | 17 | 1562 | |||
GenBank mRNA | AJ277654.1 | - | 17 | 1562 | ||
AJ277655.1 | - | 17 | 1537 | |||
AJ277656.1 | - | 17 | 1473 | |||
AJ277657.1 | - | 15 | 1348 | |||
AJ277658.1 | - | 15 | 1467 |