Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002217.1 | 6 | 1697 | |||
GenBank mRNA | AB116525.1 | - | 6 | 1697 | ||
BC041364.1 | - | 13 | 2452 |
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 15 | 2843 | ||
NM_001322003.1 | NP_001308932.1 | 15 | 2838 | |||
NM_001322004.1 | NP_001308933.1 | 14 | 2703 | |||
NM_001322005.1 | NP_001308934.1 | 15 | 2955 | |||
NM_001322006.1 | NP_001308935.1 | 14 | 2687 | |||
NM_001322007.1 | NP_001308936.1 | 14 | 2740 | |||
NM_001322008.1 | NP_001308937.1 | 13 | 2600 | |||
NM_001322009.1 | NP_001308938.1 | 15 | 2983 | |||
NM_001322010.1 | NP_001308939.1 | 13 | 2547 | |||
NM_001322011.1 | NP_001308940.1 | 15 | 2794 | |||
NM_001322012.1 | NP_001308941.1 | 15 | 2789 | |||
NM_001322013.1 | NP_001308942.1 | 14 | 2670 | |||
NM_001322014.1 | NP_001308943.1 | 15 | 2876 | |||
NM_001322015.1 | NP_001308944.1 | 15 | 3013 | |||
NR_136154.1 | 15 | 2800 | ||||
XM_006715744.3 | XP_006715807.1 | 8 | 1919 | |||
XM_017012342.1 | XP_016867831.1 | 14 | 2668 | |||
GenBank mRNA | AK125607.1 | - | 1 | 44 | ||
BQ050630.1 | - | 3 | 537 | |||
DB548617.1 | - | 1 | 67 |