Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026690.1 | 2 | 266 | |||
GenBank mRNA | CV575750.1 | - | 2 | 266 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU595969.1 | - | 300 | Chr. - - [-] on Build |