Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003064.2 | 2 | 475 | |||
GenBank mRNA | AF222904.1 | - | 2 | 224 | ||
BC093811.1 | - | 1 | 157 | |||
BC093813.1 | - | 1 | 157 |